Gatifloxacin ≥98%

Synonyms: 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

380-291-G001 380-291-G005
89150-376EA 376.56 USD
89150-376 89150-374
Gatifloxacin ≥98%
Gatifloxacin

Fluoroquinolone with broad spectrum activity against Gram-negative and Gram-positive bacteria


  • White to yellow crystalline powder


Inhibits bacterial DNA-gyrase and topoisomerase IV. May cause dysglycemia. Stimulates short-term self-renewal in both human and mouse embryonic stem cells.


Soluble in water at pH 2 - 5 (40-60 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₁₉H₂₂FN₃O₄
MW: 375.40 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00895399
CAS Number: 112811-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR