Cefotaxime sodium salt ≥95% (by HPLC)

Supplier: Enzo Life Sciences

380-271-M250 380-271-G001
89150-320EA 48.91 USD
89150-320 89150-318
Cefotaxime sodium salt ≥95% (by HPLC)
Cefotaxime sodium salt

Inhibits bacterial cell wall synthesis


  • Cephalosporin antibiotic
  • Resistant against β-lactamase


Effective against Gram-negative and Gram-positive bacteria.


White to light yellow powder


Soluble in water (>50 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₁₆H₁₇N₅O₇S₂
Storage Temperature: Freezer
MDL Number: MFCD00079073
CAS Number: 64485-93-4
https://us.vwr.com/stibo/web/std.lang.all/05/49/45390549.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR