Ampicillin sodium salt ≥93%
Supplier: Enzo Life Sciences
Synonyms:
D(-)-α-Aminobenzylpenicillin Sodium Salt
380-268-G005
89150-308EA
74.9
USD
89150-308
Ampicillin sodium salt ≥93%
Ampicillin sodium salt
β-Lactam antibiotic
- Semisynthetic from penicillin
- Inactivated by β-lactamase
Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking). Inactivates transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-negative and Gram-positive bacteria.
White to light yellow powder
Soluble in water (50 mg/mL)
Packaging: Shipping temperature: Ambient
Formula:
C₁₆H₁₈N₃NaO₄S MW: 371.39 Storage Temperature: Freezer |
MDL Number:
MFCD00064313 CAS Number: 69-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...