7-Oxostaurosporine (from Streptomyces sp. MST-AS5345) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: LCM76-LRK 1409BMY 41950

380-210-M001
89150-202EA 544.36 USD
89150-202
7-Oxostaurosporine (from Streptomyces sp. MST-AS5345) ≥98% (by HPLC)
7-Oxostaurosporine

Antibiotic. Potent inhibitor of protein kinase C (PKC). Inhibits the cell cycle at the G2 stage.


  • Apoptosis inducer. PKC inhibitor.


Yellow solid.


Soluble in DMSO, dimethyl formamide, methanol or 100% ethanol; slightly soluble in water.


PKC inhibitor.

Formula: C₂₈H₂₄N₄O₄
MW: 480.5 g/mol
Storage Temperature: Freezer
CAS Number: 125035-83-8
https://us.vwr.com/stibo/web/std.lang.all/05/13/45390513.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR