Sinefungin ≥95% (by HPLC)
Supplier: Enzo Life Sciences
380-070-M005
380-070-M001
89149-940EA
129.84
USD
89149-940
89149-942
Sinefungin ≥95% (by HPLC)
Sinefungin
Methyltransferase inhibitor
Strong competitive inhibitor of methyltransferases which uses S-adenosyl-1-methionine as the methyl group donor to yield methylated products such as 5-methylcytosine or N6-methyl adenosine on DNA and RNA. In addition, sinefungin is involved in a number of physiological processes.
CAS Number:
58944-73-3 |