Kendomycin (from Streptomyces violaceoruber) ≥98% (by HPLC)
Synonyms:
(−)-TAN 2162
380-066-C500
89149-928EA
988.54
USD
89149-928
Kendomycin (from Streptomyces violaceoruber) ≥98% (by HPLC)
Kendomycin
Yellow powder
Potent endothelin receptor antagonist. Anti-osteoporotic compound. Shows remarkable antibacterial and cytotoxic activity. Mediates its cytotoxic effects, at least in part, through proteasome inhibition.
Slightly soluble in DMSO or methanol
Packaging: Shipping temperature: Ambient
Formula:
C₂₉H₄₂O₆ MW: 486.64 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00076711 CAS Number: 183202-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...