Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Setamycin

380-063-M001
89149-918EA 361.77 USD
89149-918
Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)
Bafilomycin B1

V-ATPase inhibitor.


Macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase.


Yellow solid


Slightly soluble in methanol or DMSO, insoluble in water


Packaging: Shipping temperature: Ambient

Formula: C₄₄H₆₅NO₁₃
MW: 815.99 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00242671
CAS Number: 88899-56-3
https://us.vwr.com/stibo/web/std.lang.all/04/78/45390478.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR