Cefazolin sodium salt ≥86%
Synonyms:
Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
380-044-G001
380-044-M250
89149-888EA
248.29
USD
89149-888
89149-890
Cefazolin sodium salt ≥86%
Cefazolin sodium salt
Antibiotic
- Semisynthetic
Inhibits synthesis of bacterial cell wall.
White to off-white powder
Soluble in water (50 mg/mL), slightly soluble in methanol and 100% ethanol; insoluble in acetone, chloroform and benzene
Packaging: Shipping temperature: Ambient
Formula:
C₁₄H₁₄N₈NaO₄S₃ MW: 476.48 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00056883 CAS Number: 27164-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...