Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tautomycin ≥85% (by HPLC)

Supplier: Enzo Life Sciences

380-041-C025 380-041-C050 380-041-C100
89149-880 89149-882 89149-884
Tautomycin ≥85% (by HPLC)
Inhibitor of PP1 and PP2A.

  • Colorless film

Cell permeable protein phosphatase inhibitor

Treatment of MCF7 cells (breast caner line) with 10µM produced time-dependent inhibition of PP1 (80% in 6h, 100% in 14 h), with only 20% inhibition of PP2A after 24h.

Inhibits PP1 completely at 3nM and PP2 at 30nM. Inhibition of PP2B is over 10.000 fold weaker and PP2C is not inhibited. Structurally similar to okadaic acid but has opposite relative potency for PP1 and PP2A. Its cell permeability and unique inhibitory profile make tautomycin a useful tool for identifying phosphatase subtypes and physiological substrates for PP2 and PP2A.

Soluble in 100% ethanol, methanol or DMSO

Packaging: Shipping temperature: Ambient


Formula: C₄₁H₆₆O₁₃
MW: 767.0 g/mol
Storage Temperature: Freezer
CAS Number: 109946-35-2
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty