Oligomycin (from Streptomyces diastatochromogenes) ≥90%
Supplier: Enzo Life Sciences
Synonyms:
Oligomycin complex
, Oligomycin A, B, C
380-037-M010
380-037-M005
89149-868EA
112.84
USD
89149-868
89149-870
Oligomycin (from Streptomyces diastatochromogenes) ≥90%
Oligomycin
F-ATPase inhibitor.
Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1), preventing phosphoryl group transfer. Induces apoptosis.
White to faint yellow powder
Soluble in 100% ethanol, methanol (10 mg/mL) or acetone (50 mg/mL)
Packaging: Shipping temperature: Ambient
Formula:
C₄₅H₇₄O₁₁ MW: 789.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01779388 CAS Number: 1404-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...