Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
Supplier: Enzo Life Sciences
380-007-M001
89149-790EA
207.18
USD
89149-790
Enniatin B (from Fusarium orthoceras var. enniatum) ≥90% (by HPLC)
Enniatin B
White to off-white crystalline solid
Ionophore antibiotic. Inhibitor of the S. cerevisiae ABC transporter Pdr5p.Inhibitor of acyl-CoA-cholesterol acyltransferase and of phosphodiesterase. Potential anticancer compound.
Soluble in DMSO or other organic solvents, almost insoluble in water
Packaging: Shipping temperature: Ambient
Formula:
C₃₃H₅₇N₃O₉ Storage Temperature: Freezer |
CAS Number:
917-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...