Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cinnamtannin B-1 ≥90% (by 1H-NMR)

Supplier: Enzo Life Sciences
Synonyms: Epicatechin-(4beta→8,2beta→O→7)-epicatechin-(4alpha→8)-epicatechin

350-365-M005
Potent antioxidant

A-type proanthocyanidin contained in several plant species such as Laurus nobilis L. , Vaccinium vitis-idaea, Parameria laevigata, Cinnamomum zeylanicum, Lindera umbellata, Metaxya rostrata. Potent antioxidant, TEAC method (nM Trolox) = 0.95. Protective agent against oxidative stress and apoptosis in human platelets.
Inhibitory activity against cyclooxygenase-2 (COX-2) in vitro.
DPPH radical scavenging IC50 6.14µM
Lipid peroxidation inhibition 159.74
Nitric oxide inhibition 10.28
Antibacterial against B.subtilis (9mm, 20mg/ml)

Isolated from Laurus nobilis L.


Formula: C₄₅H₃₆O₁₈
MW: 864.7 g/mol
CAS Number: 88082-60-4


 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
5 mg 350-365-M005 89149-606 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping