(+)-Egenine (from Fumaria vaillantii or Corydalis decumbens) ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
6-(6-Methyl-5,6,7,8-tetrahydro[1,3]dioxolo[4,5-g]isoquinolin-5-yl)-6,8-dihydrofuro[3,4-E][1,3]benzodioxol-8-ol
, (+)-Decumbensine
350-362-M100
350-362-M025
89149-594EA
206.12
USD
89149-594
89149-596
(+)-Egenine (from Fumaria vaillantii or Corydalis decumbens) ≥98% (by HPLC)
(+)-Egenine
First phthalideisoquinoline hemiacetal from a natural source. Inhibits Ca2+ currents and contractile responses in vitro.
White to off-white crystalline solid.
Soluble in ethyl acetate or chloroform; sparingly soluble in 100% ethanol or ether; practically insoluble in water.
Formula:
C₂₀H₁₉NO₆ MW: 369.4 g/mol Storage Temperature: Freezer |
CAS Number:
6883-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...