Honokiol ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 3',5-Diallylbiphenyl-2,4'-diol5,3'-Diallyl-2,4'-dihydroxybiphenyl3',5-Diallyl-2,4'-dihydroxybiphenyl

350-350-M025 350-350-M005
89149-558EA 58.11 USD
89149-558 89149-560
Honokiol ≥98% (by HPLC)
Honokiol

iNOS inhibitor


Potent and highly tolerable antitumor and antiangiogenic compound. Anxiolytic, anti-thrombotic and antibacterial. Shows central depressant effects in vivo. Inhibitor of nitric oxide (NO) and TNF-alpha production in LPS-activated macrophages by the suppression of inducible nitric oxide synthase (iNOS; NOS II) expression (IC50=6.4µM). Isomeric with magnolol (Prod. No. ALX-350-352). Inhibits NF-kappaB activation. Potent scavenger of superoxide and peroxyl radicals.


Isolated from Magnolia officinalis.

Formula: C₁₈H₁₈O₂
MW: 266.34 g/mol
MDL Number: MFCD00016674
CAS Number: 35354-74-6
https://us.vwr.com/stibo/web/std.lang.all/77/56/45347756.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR