Roquefortine C ≥95% (by HPLC)

Supplier: Enzo Life Sciences

350-342-MC05
89149-540EA 555.83 USD
89149-540
Roquefortine C ≥95% (by HPLC)
Roquefortine C

Cytochrome P450 inhibitor


Potent neurotoxin produced by a diverse range of fungi, most notably Penicillium species. Inhibits growth of Gram-positive bacteria. Inhibits cytochrome p450.


Isolated from MST-FP1764.

Formula: C₂₂H₂₃N₅O₂
CAS Number: 58735-64-1
https://us.vwr.com/stibo/web/std.lang.all/77/55/45347755.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR