S14-95
Supplier: Enzo Life Sciences
350-299-MC05
89149-446EA
347.91
USD
89149-446
S14-95
S14-95
STAT1alpha inhibitor
Potent inhibitor of cytokine-induced activation of STAT1α leading to the inhibition of inducible expression of proinflammatory enzymes (COX-2, iNOS (NOS II)) and cytokines (TNF-α).
Isolated from Penicillium sp.
Formula:
C₂₈H₃₄O₅ MW: 450.57 g/mol |
CAS Number:
419532-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...