Meleagrin (from Penicillium meleagrinum) ≥98% (by HPLC)
Supplier: Enzo Life Sciences
350-297-M001
89149-438EA
285.57
USD
89149-438
Meleagrin (from Penicillium meleagrinum) ≥98% (by HPLC)
Meleagrin
Yellow oil
- Antibiotic
Isolated from
Soluble in 100% ethanol or DMSO
Packaging: Shipping temperature: Ambient
Formula:
C₂₃H₂₃N₅O₄ MW: 433.5 g/mol Storage Temperature: Freezer |
CAS Number:
71751-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...