Manzamine A ≥98% (by HPLC)
Supplier: Enzo Life Sciences
350-294-M001
89149-432EA 310.63 USD
89149-432
Manzamine A ≥98% (by HPLC)
Manzamine A
GSK-3 inhibitor
β-Carboline alkaloid with anti-inflammatory, antimalarial, antifungal, anti-HIV-1 and insecticidal activity. Also active against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus. Inhibits GSK-3. Has native fluorescence, highest when solubilized in methanol (Ex: 340nm, Em: 387nm).
Isolated from Xestospongia sp.
CAS Number: 104196-68-1
https://us.vwr.com/stibo/web/std.lang.all/77/40/45347740.jpg