Resibufogenin (from Bufonis venom) ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
(3β,5β,15β)-14,15-Epoxy-3-hydroxy-5-bufa-20,22-dienolide
350-286-M010
89149-418EA
454.45
USD
89149-418
Resibufogenin (from Bufonis venom) ≥98% (by HPLC)
Resibufogenin
Specific Na+/K+-ATPase inhibitor.
- Na+/K+ ATPase inhibitor
White to off-white solid.
Soluble in chloroform.
Formula:
C₂₄H₃₂O₄ MW: 384.5 g/mol Storage Temperature: Refrigerator |
CAS Number:
465-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...