Jasplakinolide ≥97% (by HPLC)
Supplier: Enzo Life Sciences
350-275-C050
350-275-C100
89149-390EA
513.05
USD
89149-390
89149-388
Jasplakinolide ≥97% (by HPLC)
Jasplakinolide
F-actin probe
Cell permeable F-actin probe. Induces actin polymerization. Completely inhibits the binding of phalloidin to F-actin. Antifungal and anti-proliferative agent.
Isolated from Jaspis splendens.
Formula:
C₃₆H₄₅BrN₄O₆ |
CAS Number:
102396-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...