3-ATA ≥95%
Supplier: Enzo Life Sciences
Synonyms:
3-Amino-9-thio(10H)-acridone
350-273-M005
89149-386EA
396.34
USD
89149-386
3-ATA ≥95%
3-ATA
Specific CDK4 inhibitor (IC50=3.1µM) inducing growth inhibition of p16-altered tumors.
- CDK4 inhibitor
White to off-white solid powder.
Soluble in DMSO or dimethyl formamide.
Formula:
C₁₃H₁₀N₂S MW: 226.3 g/mol Storage Temperature: Ambient |
CAS Number:
129821-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...