β-Amanitin ≥90%
350-271-M001
89149-380EA 336.3 USD
89149-380
β-Amanitin ≥90%
β-Amanitin
RNA polymerase II inhibitor
Biological activity like alpha-amanitin (Prod. No. ALX-350-270), but contains a carboxy group which is useful for coupling reactions.
Isolated from Amanita phalloides mushrooms.
CAS Number: 21150-22-1