α-Amanitin ≥90%
350-270-M001
89149-378EA 308.46 USD
89149-378
α-Amanitin ≥90%
α-Amanitin
RNA polymerase II inhibitor
Inhibits eukaryotic RNA-polymerase II with high efficiency, but not eukaryotic RNA-polymerase I, or bacterial or viral RNA-polymerases. Inhibits mammalian protein synthesis.
Isolated from Amanita phalloides mushrooms.
CAS Number: 23109-05-9