Phalloidin (FITC), Enzo Life Sciences
Supplier: Enzo Life Sciences
350-268-MC01
89149-374EA 324.85 USD
89149-374
Phalloidin (FITC), Enzo Life Sciences
Protein Linking and Labeling Systems
Actin ligand
For detection of microfilaments in the light microscope. Mixture of 4 isomers with lower affinity to actin than phalloidin (Prod. No. ALX-350-265).
Semisynthetic from aminomethyldithiolano-phalloidin.