Sceptrin dihydrochloride ≥97% (by HPLC)
350-264-M001
89149-366EA 161.56 USD
89149-366
Sceptrin dihydrochloride ≥97% (by HPLC)
Sceptrin dihydrochloride
Acetylcholine receptor inhibitor
Inhibits the growth of several Gram-positive and Gram-negative bacteria. Competitively inhibits muscarinic acetylcholine receptors.
Isolated from the sponge Agelas nakamurai.
CAS Number: 79703-25-6