Xanthorrhizol ≥97% (by HPLC)
|
Danger
|
350-263-M001
89149-364EA
189.07
USD
89149-364
Xanthorrhizol ≥97% (by HPLC)
Xanthorrhizol
Antibacterial and anti-inflammatory compound. Apoptosis inducer.
- Isolated from Curcuma xanthorrhiza.
Soluble in DMSO or 100% ethanol.
Formula:
C₁₅H₂₂O Storage Temperature: Freezer |
CAS Number:
30199-26-9 |
Specification Test Results
Purity | ≥97% (HPLC) |
Identity | Determined by ¹H-NMR, ¹³C-NMR and MS |
Source | Synthetic |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...