Triptolide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

350-259-M001 350-259-M005
89149-354EA 134.38 USD
89149-354 89149-356
Triptolide ≥98% (by HPLC)
Triptolide

Immunosuppressant


Immunosuppressive agent with anti-inflammatory and antitumor properties. More effective in preventing T cell proliferation and interferon-gamma production than FK506 (Prod. No. ALX-380-008). Induces apoptosis in T cells by activating DEVD cleaving caspases. Blocks TNF-alpha mediated induction of c-IAP1 and c-IAP2 and NF-kappaB activation. Inhibits induction of MMP-3, MMP-13, and ADAMTS-4 in cell culture.


Isolated from Tripterygium wilfordii.

Formula: C₂₀H₂₄O₆
MDL Number: MFCD00210565
CAS Number: 38748-32-2
https://us.vwr.com/stibo/web/std.lang.all/77/24/45347724.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR