Sedanolide ≥98% (by HPLC)
Supplier: Enzo Life Sciences
350-229-M100
89149-292EA
215.23
USD
89149-292
Sedanolide ≥98% (by HPLC)
Sedanolide
GST activator
Inducer of glutathione S-transferases and inhibitor of chemically induced carcinogenesis. Mosquitocidal, nematicidal and antifungal. Was shown to inhibit cyclooxygenase-1 and -2 (COX-1 and COX-2) as well as topoisomerase I and topoisomerase II.
Isolated from Apium graveolens L.
CAS Number:
6415-59-4 |