Kahweol ≥97% (by UPLC)
Supplier: Enzo Life Sciences
350-223-M010
89149-282EA
160.51
USD
89149-282
Kahweol ≥97% (by UPLC)
Kahweol
GST activator.
Induces glutathione S-transferase. Has anti-cancer, chemoprotective, anti-inflammatory and anti-atherosclerotic properties.
Soluble in ethyl acetate, acetone or DMSO; insoluble in water.
Formula:
C₂₀H₂₆O₃ Storage Temperature: Freezer |
CAS Number:
6894-43-5 |
Specification Test Results
Purity | ≥97% (UPLC) |
Source | Natural product isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...