Cafestol ≥98% (by UHPLC)
Supplier: Enzo Life Sciences
350-220-M050
89149-278EA
256.65
USD
89149-278
Cafestol ≥98% (by UHPLC)
Cafestol
GST activator.
Inducer of glutathione S-transferases. Has chemoprotective activity, reducing the genotoxicity of several carcinogens.
Soluble in DMSO, 100% ethanol or ethyl acetate.
Formula:
C₂₀H₂₈O₃ MW: 316.441 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01075769 CAS Number: 469-83-0 |
Specification Test Results
Purity | ≥98% (UHPLC) |
Identity | Determined by ¹H-NMR |
Source | Isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...