(-)-Eburnamonine ≥99%
Supplier: Enzo Life Sciences
Synonyms:
(3α,16α)-Eburnamenin-14(15H)-one
, vinburnine
, vincamone
350-216-M100
89149-272EA
133.71
USD
89149-272
(-)-Eburnamonine ≥99%
(-)-Eburnamonine
Antihypoxic. Vasodilator.
Cerebral metabolic stimulant; shows antihypoxic effects.
Soluble in chloroform.
Synonym: (3α,16α)-Eburnamenin-14(15H)-one, vincamone, vinburnine
Formula:
C₁₉H₂₂N₂O MW: 294.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00064309 CAS Number: 4880-88-0 EINECS: 225-490-5 |
Specification Test Results
Purity | ≥99% (Assay) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...