Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[D-Asp3]microcystin-LR, Enzo Life Sciences

Supplier: Enzo Life Sciences
350-173-C025 350-173-C100
Microcystins are a group of cyclic heptapeptide hepatotoxins produced by a number of cyanobacterial genera. The most notable of which, and namesake, is the widespread genus Microcystis. Structurally, all microcystins consist of the generalized structure cyclo(-D-Ala1-X2-D-MeAsp3-Y4-Adda5-D-Glu6-Mdha7-). X and Y are variable L-amino acids, D-MeAsp is D-erythro-β-methylaspartic acid and Mdha is N-methyldehydroalanine. Adda is the cyanobacteria unique C20 β-amino acid 3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-deca-4,6-dienoic acid. Substitutions of the variable L-amino acids at positions 2 and 4 give rise to at least 21 known primary microcystin analogs and alterations in the other constituent amino acids result in more than 90 reported mycrocystins to date.

Isolated from Microcystis aeruginosa. Microcystin containing a leucine (L) in position 2 and arginine (R) in position 4. Additionally, the D-MeAsp at position 3 is demethylated to Asp. Hepatotoxic.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 350-173-C100 89149-250 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 µg 350-173-C025 89149-248 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name [D-Asp3]microcystin-LR
Species Microcystis aeruginosa
Purity ≥95% (HPLC)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping