Neobavaisoflavone ≥97% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
7-Hydroxy-3-[4-hydroxy-3-(3-methyl-2-buten-1-yl)phenyl]-4H-1-benzopyran-4-one
, 7-Hydroxy-3-(4-hydroxy-3-(3-methylbut-2-en-1-yl)phenyl)-4H-chromen-4-one
, 4',7-Dihydroxy-3'-(3-methyl-2-butenyl)isoflavone
350-146-M001
89149-168EA
271.28
USD
89149-168
Neobavaisoflavone ≥97% (by HPLC)
Neobavaisoflavone
Inhibits platelet aggregation. DNA polymerase inhibitor. Shows antifungal activity.
- DNA polymerase inhibitor
Yellow solid.
Soluble in DMSO or 100% ethanol.
Formula:
C₂₀H₁₈O₄ MW: 322.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00800660 CAS Number: 41060-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...