Tentoxin (from Alternaria alternata) ≥98% (by HPLC, TLC)

Supplier: Enzo Life Sciences

Synonyms: cyclo-(N-Methyl-L-alanyl-L-leucyl-N-methyl-(Z)-dihydrophenylalanyl-glycyl)

350-132-MC05
89149-136EA 302.56 USD
89149-136
Tentoxin (from Alternaria alternata) ≥98% (by HPLC, TLC)
Tentoxin

Cyclic tetrapeptide. Inhibitor of chloroplast F1-ATPase (CF1). Does not inhibit the homologous enzymes from chloroplasts of insusceptible plant species or from bacteria or mitochondria.


  • F-ATPase inhibitor


Off-white to yellow powder.


Soluble in DMSO (10mg/ml) or methanol (10mg/ml).

Formula: C₂₂H₃₀N₄O₄
MW: 414.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 28540-82-1
https://us.vwr.com/stibo/web/std.lang.all/76/81/45347681.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR