Aristoforin ≥95%

350-129-M001
89149-130EA 352.47 USD
89149-130
Aristoforin ≥95%
Aristoforin

Stable and water-soluble derivative of hyperforin inducing apoptosis. Antitumor agent. Inhibits sirtuins.


  • SIRT inhibitor


White to yellow oil.


Soluble in DMSO or 100mM sodium hydrogen carbonate/2% DMSO.

Formula: C₃₇H₅₄O₆
MW: 594.8 g/mol
Storage Temperature: Freezer
CAS Number: 849215-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR