Chetomin ≥98% (by HPLC)
350-128-M001 350-128-M005
89149-126EA 253.78 USD
89149-126 89149-128
Chetomin ≥98% (by HPLC)
Chetomin
HIF-1 inhibitor
Dithiodiketopiperazine inhibitor of HIF-1 formation by disrupting the binding of p300 to both HIF-1α and HIF-2α. Inhibitor of tumor growth. Potent immunosuppressor. Antibacterial.
Isolated from Chaetomium species.
CAS Number: 1403-36-7