Polydatin ≥98% (by HPLC)

Synonyms: (2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-(4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

350-114-M010
89149-094EA 144.95 USD
89149-094
Polydatin ≥98% (by HPLC)
Polydatin

Platelet aggregation inhibitor


Platelet aggregation inhibitor. Shows multiple effects on vascular smooth muscle cells, myocardial cells, endothelial cells and white blood cells after shock. May inhibit phospholipase A2. Inhibits the expression of various cell adhesion molecules.


Isolated from Polygonum cuspidatum.

Formula: C₂₀H₂₂O₈
MW: 390.3839 g/mol
MDL Number: MFCD00210592
CAS Number: 65914-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR