Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferutinin ≥98% (by HPLC), white to off-white solid, high purity

Supplier: Enzo Life Sciences

350-098-M005 350-098-M010
Estrogen receptor α activator.

Potent, naturally occuring non-steroid estrogenic compound. Agonist for estrogen receptor (ER) α and agonist/antagonist for ERβ with higher binding affinity than tamoxifen for both ERs. Electrogenic Ca²⁺ ionophore inducing mitochondrial depolarization which can be completely blocked by cyclosporin A, suggesting that ferutinin opens the mitochondrial permeability transition pore (mPTP). In a concentration range of 1 - 50 µM ferutinin increases the permeability of thymocytes, mitochondria, sarcoplasmic reticulum, liposomes and bilayer lipid membranes for Ca²⁺.

Soluble in acetone, dichloromethane, DMSO or ethyl acetate.


Formula: C₂₂H₃₀O₄
MW: 358,5
Storage Temperature: Refrigerator
CAS Number: 41743-44-6
Specification Test Results
Purity ≥98% by HPLC
Source Semisynthetic


 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
5 mg 350-098-M005 89149-054 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

10 mg 350-098-M010 89149-056 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping