Sanguinarine chloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
Pseudochelerythrine chloride
, Pseudochelerythrine
350-076-M005
89149-016EA
229.83
USD
89149-016
Sanguinarine chloride ≥98% (by HPLC)
Sanguinarine chloride
Na+/K+- and Mg2+-ATPase inhibitor
Na+/K+-and Mg2+-ATPase inhibitor. Antimicrobial, anti-inflammatory and antioxidant properties. Induces apoptosis. Modulates the Bcl-2 family. Shows antiplaque activity in humans.
Isolated from the leaves and stems of Macleaya cordata and M. microcarpa.
Formula:
C₂₀H₁₄NO₄Cl MW: 367.78 g/mol |
MDL Number:
MFCD00012126 CAS Number: 5578-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...