Would you like to log in again?
Olomoucine (high purity), Enzo Life Sciences
Supplier: Enzo Life Sciences
350-013-M025 350-013-M005
CDK/cyclin inhibitor
Olomoucine is a purine derivative which acts as a potent and selective inhibitor of CDK1 (p34cdc2)/cyclin B and related kinases. It acts by competing for the ATP binding domain of the kinase. Of 35 protein kinases tested, olomoucine inhibited p33CDK2/cyclin A, p33CDK2/cyclin E, p34cdc2/cyclin B, p33CDK5/p35 and erk1 with IC50 of 7, 7, 7, 3 and 25µM respectively. The related kinases p34CDK4/cyclin D and p40CDK6/cyclin D3 were not inhibited or were less sensitive to olomoucine, IC50 > 1000 and 250µM respectively. PKC, PKA, PKG, CaM kinase II, MLC kinase, CK2, EGFR tyrosine kinase, p56lck, p56lyn, p60c-src, v-abl and insulin receptor tyrosine kinase are not inhibited or are weakly inhibited at 1mM. It delays mouse oocyte maturation and inhibits the growth of a variety of human cell lines including Rhabdomyosarcoma cell line Rh1, Rh18, Rh28 and Rh30; MCF-7, KB-3-1 and their adriamycin-resistant counterparts (IC50=18-86µM). It inhibits DNA synthesis in interleukin-2-stimulated T lymphocytes and triggers a G1 arrest similar to interleukin-2 deprivation. Can be used to synchronize cells in G1. Triggers apoptosis in target tumor cells in vitro and in vivo and in maturing cerebellar granule neurons. N9-Isopropyl-olomoucine and the negative control Iso-olomoucine are also available.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 mg 350-013-M025 89148-934 Each Retrieving Restricted
Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.
5 mg 350-013-M005 89148-932 Each Retrieving Restricted
Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Purity ≥98%
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty