Dynasore ≥98% (by NMR)

Supplier: Enzo Life Sciences

Synonyms: 3-Hydroxynaphthalene-2-carboxylic acid (3,4-Dihydroxybenzylidene)hydrazideN'-(3,4-Dihydroxybenzylidene)-3-hydroxy-2-naphthohydrazide

270-502-M005 270-502-M025
89148-114EA 74.9 USD
89148-114 89148-116
Dynasore ≥98% (by NMR)
Dynasore

GTPase inhibitor


Small molecule GTPase inhibitor targeting Dynamin-1, Dynamin-2 and Drp1 (mitochondrial Dynamin). Noncompetitive inhibitor of Dynamin GTPase activity. Blocks Dynamin-dependent endocytosis in cells (including neurons). Fast acting (seconds). Strong, dose dependent block of the traffic and accumulation of transferrin with 80µM dynasore (IC50 = 15µM). Has also been shown to block human papillomavirus type 16 (HPV16) and bovine papillomavirus type 1 (BPV1) (pseudovirions) infections.

Formula: C₁₈H₁₄N₂O₄
MW: 322.32 g/mol
MDL Number: MFCD00292551
CAS Number: 304448-55-3
https://us.vwr.com/stibo/web/std.lang.all/76/07/45347607.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR