Fenofibrate ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate
, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester
PPARα activator.
Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist (EC₅₀ = 18 µM for mouse and EC₅₀ = 30 µM for human PPARα). Also binds to PPARγ, but with at least 10-fold less affinity and is inactive at PPARδ (up to EC₅₀ = 100 µM).
Soluble in acetone, ether, DMSO or dimethyl formamide; slightly soluble in 100% ethanol or methanol; insoluble in water.
Synonyms: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methyl-propanoic acid 1-methylethyl ester, procetofen
Formula:
C₂₀H₂₁ClO₄ MW: 360.83 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00133314 CAS Number: 49562-28-9 EINECS: 256-376-3 |
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...