Ciprofibrate ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
2-(4-(2,2-Dichlorocyclopropyl)phenoxy)-2-methylpropanoic acid
, 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid
270-475-M100
270-475-M025
89148-026EA
72.45
USD
89148-026
89148-028
Ciprofibrate ≥98% (by HPLC)
Ciprofibrate
PPARα activator.
Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist.
Soluble in DMSO.
Synonym: 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid
Formula:
C₁₃H₁₄Cl₂O₃ MW: 289.15 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00467135 CAS Number: 52214-84-3 EINECS: 257-744-6 |
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...