AS-605240
Supplier: Enzo Life Sciences
Synonyms:
5-(Quinoxalin-6-ylmethylene)thiazolidine-2,4-dione
270-462-M005
89147-978EA
118.08
USD
89147-978
AS-605240
AS-605240
Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ)
Exhibits selectivity over other PI(3)K isoforms.
Formula:
C₁₂H₇N₃O₂S MW: 257.2679 g/mol |
MDL Number:
MFCD11114066 CAS Number: 648450-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...