Sphingolactone-24 ≥95%

Supplier: Enzo Life Sciences

270-453-M001
89147-948EA 305.4 USD
89147-948
Sphingolactone-24 ≥95%
Sphingolactone-24

Selective and irreversible inhibitor of neutral sphingomyelinase (N-SMase).


  • Sphingomyelinase inhibitor


White waxy solid.


Soluble in DMSO (10mg/ml).

Formula: C₁₈H₂₉NO₄
MW: 323.4 g/mol
Storage Temperature: Freezer
CAS Number: 881177-99-7
https://us.vwr.com/stibo/web/std.lang.all/75/77/45347577.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR