DR2313
Supplier: Enzo Life Sciences
Synonyms:
7,8-Dihydro-2-methyl-1H-thiopyrano[4,3-d]pyrimidin-4(5H)-one
270-452-M005
270-452-M025
89147-946EA
258.76
USD
89147-946
89147-944
DR2313
DR2313
PARP inhibitor
Potent, water soluble competitive PARP inhibitor (IC50=0.20µM and 0.24µM for PARP-1 and PARP-2 respectively).
Formula:
C₈H₁₀N₂OS MW: 182.2428 g/mol |
MDL Number:
MFCD08703132 CAS Number: 284028-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...