Trans-24 ≥95%

Synonyms: (5R,11aS)-2-Benzyl-5-(3-hydroxyphenyl)-6H-1,2,3,5,11,11a-hexahydroimidazo[1,5-b]-β-carboline-1,3-dione

270-439-M001
89147-904EA 147.06 USD
89147-904
Trans-24 ≥95%
Trans-24

Potent, cell permeable non-tubulin-interacting mitosis inhibitor. Blocks mitosis (IC50=650nM) by binding to the mitotic kinesin Eg5.


  • Mitotic kinesin Eg5 inhibitor


Yellow solid.


Soluble in DMSO (10mg/ml).

Formula: C₂₆H₂₁N₃O₃
MW: 423.5 g/mol
Storage Temperature: Freezer
CAS Number: 869304-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR