Dimethylenastron ≥95%
Synonyms:
7,7-Dimethyl-4-(3-hydroxyphenyl)-5-oxo-3,4,5,6,7,8-hexahydroquinazolin-2(1H)-thione
270-438-M001
89147-902EA
167.59
USD
89147-902
Dimethylenastron ≥95%
Dimethylenastron
Potent, cell permeable non-tubulin-interacting mitosis inhibitor. Blocks mitosis (IC50=200nM) by binding to the mitotic kinesin Eg5.
- Mitotic kinesin Eg5 inhibitor
Yellow solid.
Soluble in DMSO (10mg/ml) or methanol (5mg/ml).
Formula:
C₁₆H₁₈N₂O₂S MW: 302.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09264157 CAS Number: 863774-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...