1,1-Dimethylbiguanide hydrochloride ≥97%

Supplier: Enzo Life Sciences

Synonyms: AMPK Signaling Activator VIIMetformin1,1-Dimethylbiguanide HydrochlorideMetformin hydrochloride

270-432-G005
89147-892EA 50.86 USD
89147-892
1,1-Dimethylbiguanide hydrochloride ≥97%
1,1-Dimethylbiguanide hydrochloride

Antidiabetic agent


Metformin is an antidiabetic agent. Reduces the cardiovascular complications of diabetes. Does not induce hypoglycemia. Inhibits hepatic gluconeogenesis via activation of the LKB1/AMPK pathway. Has antiproliferative effects in cancer cell lines.

Formula: C₄H₁₂ClN₅
MW: 165.625 g/mol
MDL Number: MFCD00012582
CAS Number: 1115-70-4
https://us.vwr.com/stibo/web/std.lang.all/75/67/45347567.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR