SC-560 ≥98%

Supplier: Enzo Life Sciences

Synonyms: 5-(4-Chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluoromethyl)-1H-pyrazole

270-427-M025 270-427-M005
89147-872EA 90.18 USD
89147-872 89147-874
SC-560 ≥98%
SC-560

Selective COX-1 inhibitor (IC50=9nM). Shows 700-fold more selectivity over COX-2 (IC50=6.3µM). Inhibits COX-1-derived platelet thromboxane B2, gastric PGE2, and dermal PGE2 production. Orally active in rat.


  • COX inhibitor


White to off-white crystalline solid.


Soluble in 100% ethanol, DMSO or dimethyl formamide; sparingly soluble in aqueous buffers.

Formula: C₁₇H₁₂N₂OClF₃
MW: 352.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02179214
CAS Number: 188817-13-2
https://us.vwr.com/stibo/web/std.lang.all/75/63/45347563.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR